missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel(II) oxalate dihydrate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03945430

 View more versions of this product

Catalog No. AA3945430

Explore available promotions

Description

Description

It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nickel(II) oxalate dihydrate
Odorless
MFCD00167155
UN3077
nickel oxalate, nickel ii oxalate dihydrate, nickel oxalate dihydrate, acmc-1b92x, nickel 2+ ; oxalate; dihydrate, nickel oxalate,, nickel 2+ dihydrate oxalate, nickel 2+ ion dihydrate oxalate
VNYOIRCILMCTHO-UHFFFAOYSA-L
nickel(2+);oxalate;dihydrate
516789
Powder
6018-94-6
C2H4NiO6
250 g
14,6514
Soluble in acids; Insoluble in water
C(=O)(C(=O)[O-])[O-].O.O.[Ni+2]
182.741
182.76 (146.73 Anhydrous)

Chemical Identifiers

6018-94-6
182.741
VNYOIRCILMCTHO-UHFFFAOYSA-L
516789
C(=O)(C(=O)[O-])[O-].O.O.[Ni+2]
C2H4NiO6
MFCD00167155
nickel oxalate, nickel ii oxalate dihydrate, nickel oxalate dihydrate, acmc-1b92x, nickel 2+ ; oxalate; dihydrate, nickel oxalate,, nickel 2+ dihydrate oxalate, nickel 2+ ion dihydrate oxalate
nickel(2+);oxalate;dihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P312-P330-P342+P311-P363-P501c

H302+H312-H334-H350-H372

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 208-933-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only