Learn More
Nickel(II) perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01163136
Description
Nickel(II) perchlorate hexahydrate is used as a starting material for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel(II) perchlorate hexahydrate | |
13520-61-1 | |
Damp Crystalline Solid | |
Cl2H12NiO14 | |
nickel ii perchlorate hexahydrate, acmc-1boln, 2clo4.ni.6h2o, nickel ii perchlorate hydrate, nickel 2+ hexahydrate diperchlorate, nickel perchlorate, hexahydrate, reagent, nickel ii perchlorate hexahydrate, reagent grade, nickel ii perchlorate hexahydrate trace metals basis, nickel ii perchlorate hexahydrate, purum p.a., crystallized kt | |
WHGYCGOFTBFDLW-UHFFFAOYSA-L | |
nickel(2+);diperchlorate;hexahydrate | |
15769463 | |
Reagent | |
1.55 | |
Green crystals |
140°C | |
Green | |
500 g | |
MFCD00149804 | |
Very soluble in water. Soluble in many organic solvents | |
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ni+2] | |
365.675 | |
365.70 (257.59 Anhydrous) | |
Odorless | |
Hygroscopic |
Chemical Identifiers
13520-61-1 | |
365.675 | |
WHGYCGOFTBFDLW-UHFFFAOYSA-L | |
15769463 | |
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ni+2] |
Cl2H12NiO14 | |
MFCD00149804 | |
nickel ii perchlorate hexahydrate, acmc-1boln, 2clo4.ni.6h2o, nickel ii perchlorate hydrate, nickel 2+ hexahydrate diperchlorate, nickel perchlorate, hexahydrate, reagent, nickel ii perchlorate hexahydrate, reagent grade, nickel ii perchlorate hexahydrate trace metals basis, nickel ii perchlorate hexahydrate, purum p.a., crystallized kt | |
nickel(2+);diperchlorate;hexahydrate |
Safety and Handling
Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
P201-P202-P210-P220-P221-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P370+P378q-P501c
H272-H314-H317-H334-H335-H341-H350-H360-H372
DOTInformation : DOT Class: 5.1, Packaging Group: II
EINECSNumber : 237-124-1
RTECSNumber : SC9550000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only