Learn More
Nickel(II) sulfamate hydrate, Thermo Scientific Chemicals
$106.29 - $1124.93
Chemical Identifiers
CAS | 13770-89-3 |
---|---|
Molecular Formula | H4N2NiO6S2 |
Molecular Weight (g/mol) | 250.85 |
MDL Number | MFCD00137261 |
InChI Key | KERTUBUCQCSNJU-UHFFFAOYSA-L |
Synonym | nickel sulfamate, nickel bis sulphamidate, nickel ii sulfamate, aeronikl 250, aeronikl 400, aeronikl 575, nickel 2+ disulfamate, sulfamic acid, nickel 2+ salt 2:1, nickel sulfamate 6ci,7ci |
PubChem CID | 83720 |
SMILES | [Ni++].NS([O-])(=O)=O.NS([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AA8902018
|
Thermo Scientific Chemicals
08902018 |
50 g |
Each for $106.29
|
|
||||||
AA8902030
|
Thermo Scientific Chemicals
08902030 |
250 g |
Each for $343.32
|
|
||||||
AA89020A1
|
Thermo Scientific Chemicals
089020A1 |
1 kg |
Each for $1,124.93
|
|
||||||
Description
Nickel(II) sulfamate is used as an electro plating and surface treating agent. It finds application in metal coloring and casting.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13770-89-3 | |
250.85 | |
KERTUBUCQCSNJU-UHFFFAOYSA-L | |
83720 |
H4N2NiO6S2 | |
MFCD00137261 | |
nickel sulfamate, nickel bis sulphamidate, nickel ii sulfamate, aeronikl 250, aeronikl 400, aeronikl 575, nickel 2+ disulfamate, sulfamic acid, nickel 2+ salt 2:1, nickel sulfamate 6ci,7ci | |
[Ni++].NS([O-])(=O)=O.NS([O-])(=O)=O |
Specifications
Blue | |
Crystals | |
H4N2NiO6S2 | |
nickel sulfamate, nickel bis sulphamidate, nickel ii sulfamate, aeronikl 250, aeronikl 400, aeronikl 575, nickel 2+ disulfamate, sulfamic acid, nickel 2+ salt 2:1, nickel sulfamate 6ci,7ci | |
KERTUBUCQCSNJU-UHFFFAOYSA-L | |
nickel(2+) disulfamate | |
83720 | |
Odorless |
13770-89-3 | |
50 g | |
MFCD00137261 | |
Soluble in water. | |
[Ni++].NS([O-])(=O)=O.NS([O-])(=O)=O | |
250.85 | |
250.86 (Anhydrous) | |
Nickel(II) sulfamate hydrate |
Safety and Handling
P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H341-H350i-H360D-H372
DOTInformation : DOT Class: 9, Packaging Group: III
EINECSNumber : 237-396-1
RTECSNumber : QR9275000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only