missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel(II) sulfate heptahydrate, for analysis, for nickel plating, DIN 50970, Thermo Scientific Chemicals

$153.53 - $430.27

Chemical Identifiers

CAS 10101-98-1
Molecular Formula H14NiO11S
Molecular Weight (g/mol) 280.85
MDL Number MFCD00149814
InChI Key OGKAGKFVPCOHQW-UHFFFAOYSA-L
Synonym nickel ii sulfate heptahydrate, nickel sulfate heptahydrate, nickel 2+ sulfate heptahydrate, nickelous sulfate heptahydrate, unii-596idd57nr, sulfuric acid, nickel 2+ salt 1:1 , heptahydrate, nickel sulfate heptahydrate nickel and nickel compounds, ni.so4.7h2o, ksc492q3n, nickel sulfate-water 1/7
PubChem CID 61474
ChEBI CHEBI:53504
SMILES O.O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC270552500
SDS View Documents
Thermo Scientific Chemicals
270552500
250 g Plastic bottle
Each for $153.53
 
AC270550010
SDS View Documents
Thermo Scientific Chemicals
270550010
1 kg Plastic bottle
Each for $430.27
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10101-98-1
280.85
OGKAGKFVPCOHQW-UHFFFAOYSA-L
61474
O.O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O
H14NiO11S
MFCD00149814
nickel ii sulfate heptahydrate, nickel sulfate heptahydrate, nickel 2+ sulfate heptahydrate, nickelous sulfate heptahydrate, unii-596idd57nr, sulfuric acid, nickel 2+ salt 1:1 , heptahydrate, nickel sulfate heptahydrate nickel and nickel compounds, ni.so4.7h2o, ksc492q3n, nickel sulfate-water 1/7
CHEBI:53504

Specifications

10101-98-1,7786-81-4
Needle-like Crystals or Powder
H14NiO11S
MFCD00149814
nickel ii sulfate heptahydrate, nickel sulfate heptahydrate, nickel 2+ sulfate heptahydrate, nickelous sulfate heptahydrate, unii-596idd57nr, sulfuric acid, nickel 2+ salt 1:1 , heptahydrate, nickel sulfate heptahydrate nickel and nickel compounds, ni.so4.7h2o, ksc492q3n, nickel sulfate-water 1/7
OGKAGKFVPCOHQW-UHFFFAOYSA-L
nickel(2+);sulfate;heptahydrate
61474
280.88
Analytical
Nickel(II) sulfate heptahydrate
Green
250 g
NiSO4·7H2O
15, 6602
Solubility in water: 625g/L (20°C). Other solubilities: insoluble in alcohol and ether
O.O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O
280.85
CHEBI:53504
20 to 21% (Ni)
Plastic bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Suspected of causing genetic defects.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
M

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only