missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel(II) sulfate hexahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 415611000

 View more versions of this product

Catalog No. AC415611000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nickel(II) sulfate hexahydrate
10101-97-0,7786-81-4
98.0
Crystals
98.0 to 102% (Complexometry)
NiSO4·6H2O
15,6602
Solubility in water: 625g/L (20°C). Other solubilities: sparingly soluble in alcohol
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
262.85
CHEBI:53437
≥98%
Plastic bottle
53.0°C
Blue-Green
100.0
100 g
NiO46H2O
MFCD00149813
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
RRIWRJBSCGCBID-UHFFFAOYSA-L
nickel(2+);sulfate;hexahydrate
5284429
262.85
ACS Reagent

Chemical Identifiers

10101-97-0
262.85
RRIWRJBSCGCBID-UHFFFAOYSA-L
5284429
nickel(2+);sulfate;hexahydrate
NiO46H2O
MFCD00149813
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
CHEBI:53437
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Suspected of causing genetic defects.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
M

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
IF exposed or concerned: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.

GHS Signal Word: Danger

RTECSNumber : QR9600000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only