Learn More
Nickel(II) sulfate hydrate, Puratronic™, 99.9985% (metals basis)
CAS: 15244-37-8 | NiO4S | 154.749 g/mol
$180.05 - $575.49
Chemical Identifiers
| CAS | 15244-37-8 |
|---|---|
| Molecular Formula | NiO4S |
| Molecular Weight (g/mol) | 154.749 |
| MDL Number | MFCD00011146 |
| InChI Key | LGQLOGILCSXPEA-UHFFFAOYSA-L |
| Synonym | nickel sulfate, nickel sulphate, nickel ii sulfate, nickelous sulfate, nickel 2+ sulfate, nickel sulfate 1:1, niso4, nickel ii sulfate 1:1, nickel sulfate van, nickel sulfate niso4 |
| PubChem CID | 24586 |
| ChEBI | CHEBI:53001 |
| IUPAC Name | nickel(2+);sulfate |
| SMILES | [O-]S(=O)(=O)[O-].[Ni+2] |
Description
Nickel(II) sulfate hydrate is used in electroplating and as a chemical intermediate to produce other nickel compounds. It is also used in printing and in dyeing of textiles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 15244-37-8 | |
| 154.749 | |
| LGQLOGILCSXPEA-UHFFFAOYSA-L | |
| 24586 | |
| nickel(2+);sulfate |
| NiO4S | |
| MFCD00011146 | |
| nickel sulfate, nickel sulphate, nickel ii sulfate, nickelous sulfate, nickel 2+ sulfate, nickel sulfate 1:1, niso4, nickel ii sulfate 1:1, nickel sulfate van, nickel sulfate niso4 | |
| CHEBI:53001 | |
| [O-]S(=O)(=O)[O-].[Ni+2] |
Specifications
| 15244-37-8 | |
| 4.3 to 4.7 | |
| 25 g | |
| NiSO4·xH2O (x≈6) | |
| UN3288 | |
| nickel sulfate, nickel sulphate, nickel ii sulfate, nickelous sulfate, nickel 2+ sulfate, nickel sulfate 1:1, niso4, nickel ii sulfate 1:1, nickel sulfate van, nickel sulfate niso4 | |
| LGQLOGILCSXPEA-UHFFFAOYSA-L | |
| nickel(2+);sulfate | |
| 24586 | |
| 154.77 (Anhydrous) | |
| Puratronic | |
| 2.070 g/mL | |
| (metals basis) |
| Green | |
| Crystalline Powder | |
| NiO4S | |
| MFCD00011146 | |
| 14,6517 | |
| Soluble in water (29.3g/100cc water at 0°C and 83.7g/100cc water at 100°C. Insoluble in alcohol,ether & acetone. | |
| [O-]S(=O)(=O)[O-].[Ni+2] | |
| 154.749 | |
| CHEBI:53001 | |
| 99.9985% | |
| Odorless | |
| Nickel(II) sulfate hydrate |
Safety and Handling
GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
H302+H332-H315-H317-H319-H334-H341-H350i-H360D-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber : 232-104-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only