missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nickel Nitrate, Crystal, Spectrum™ Chemical
$189.32 - $12308.41
Chemical Identifiers
CAS | 13478-00-7 |
---|---|
Molecular Formula | H12N2NiO12 |
Molecular Weight (g/mol) | 290.79 |
MDL Number | MFCD00149805 |
InChI Key | AOPCKOPZYFFEDA-UHFFFAOYSA-N |
IUPAC Name | nickel(2+) hexahydrate dinitrate |
SMILES | O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18611971
|
Spectrum Chemical Mfg Cor
N106225GM |
25 g | Amber Glass Bottle |
Each for $189.32
|
|
||||
18611966
|
Spectrum Chemical Mfg Cor
N1062125GM |
125 g | Amber Glass Bottle |
Each for $341.18
|
|
||||
18611967
|
Spectrum Chemical Mfg Cor
N106212KG |
12 kg | Poly Pail |
Each for $4,710.09
|
|
||||
18611970
|
Spectrum Chemical Mfg Cor
N106245KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
13478-00-7 | |
290.79 | |
AOPCKOPZYFFEDA-UHFFFAOYSA-N | |
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
H12N2NiO12 | |
MFCD00149805 | |
nickel(2+) hexahydrate dinitrate |
Specifications
13478-00-7 | |
25 g | |
MFCD00149805 | |
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
290.79 | |
Amber Glass Bottle |
1 | |
H12N2NiO12 | |
AOPCKOPZYFFEDA-UHFFFAOYSA-N | |
nickel(2+) hexahydrate dinitrate | |
Reagent |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.