missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel tin oxide dihydrate, Thermo Scientific Chemicals

$200.88 - $969.70

Chemical Identifiers

CAS 12035-38-0
Molecular Formula NiO3Sn
Molecular Weight (g/mol) 225.40
MDL Number MFCD00054011
InChI Key SHULAURIUQUJKN-UHFFFAOYSA-N
Synonym Nickel stannate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3945222
View Documents
Thermo Scientific Chemicals
03945222
100 g
Each for $200.88
 
AA3945236
View Documents
Thermo Scientific Chemicals
03945236
500 g
Each for $969.70
 
Description

Description

It is used as an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12035-38-0
225.40
SHULAURIUQUJKN-UHFFFAOYSA-N
NiO3Sn
MFCD00054011
Nickel stannate

Specifications

Green
Powder
NiO3Sn
Nickel stannate
SHULAURIUQUJKN-UHFFFAOYSA-N
261.43 (225.40 Anhydrous)
Nickel tin oxide dihydrate
12035-38-0
100 g
MFCD00054011
Insoluble in water.
225.40
Odorless
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H350i-H372

EINECSNumber : 234-824-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only