missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nicorandil, 98+%, Thermo Scientific Chemicals

A vasodilator that activates ATP-sensitive potassium channels

Supplier:  Thermo Scientific Chemicals J60879MD

 View more versions of this product

Catalog No. AAJ60879MD


Explore available promotions

Description

Description

A vasodilator that activates ATP-sensitive potassium channels

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nicorandil
92°C to 93°C
C8H9N3O4
250 mg
nicorandil, ikorel, 2-nicotinamidoethyl nitrate, adancor, dancor, nicorandilum, sigmart, 2-pyridine-3-carbonylamino ethyl nitrate, n-2-hydroxyethyl nicotinamide nitrate, nicorandilum inn-latin
[O-][N+](=O)OCCNC(=O)C1=CC=CN=C1
211.18
CHEBI:31905
≥98%
65141-46-0
White
MFCD00186520
14,6521
LBHIOVVIQHSOQN-UHFFFAOYSA-N
2-[(pyridin-3-yl)formamido]ethyl nitrate
47528
211.17
Powder

Chemical Identifiers

65141-46-0
211.18
LBHIOVVIQHSOQN-UHFFFAOYSA-N
47528
2-[(pyridin-3-yl)formamido]ethyl nitrate
C8H9N3O4
MFCD00186520
nicorandil, ikorel, 2-nicotinamidoethyl nitrate, adancor, dancor, nicorandilum, sigmart, 2-pyridine-3-carbonylamino ethyl nitrate, n-2-hydroxyethyl nicotinamide nitrate, nicorandilum inn-latin
CHEBI:31905
[O-][N+](=O)OCCNC(=O)C1=CC=CN=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.

P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c

H302-H318

EINECSNumber : 265-514-1

RTECSNumber : US4667600

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only