missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nicotinamide 99.0+%, TCI America™
$39.51 - $108.62
Chemical Identifiers
CAS | 98-92-0 |
---|---|
Molecular Formula | C6H6N2O |
Molecular Weight (g/mol) | 122.127 |
MDL Number | MFCD00006395 |
InChI Key | DFPAKSUCGFBDDF-UHFFFAOYSA-N |
Synonym | nicotinamide, niacinamide, 3-pyridinecarboxamide, vitamin pp, nicotinic acid amide, papulex, aminicotin, amixicotyn, nicobion, nicotylamide |
PubChem CID | 936 |
ChEBI | CHEBI:17154 |
IUPAC Name | pyridine-3-carboxamide |
SMILES | C1=CC(=CN=C1)C(=O)N |
Chemical Identifiers
98-92-0 | |
122.127 | |
DFPAKSUCGFBDDF-UHFFFAOYSA-N | |
936 | |
pyridine-3-carboxamide |
C6H6N2O | |
MFCD00006395 | |
nicotinamide, niacinamide, 3-pyridinecarboxamide, vitamin pp, nicotinic acid amide, papulex, aminicotin, amixicotyn, nicobion, nicotylamide | |
CHEBI:17154 | |
C1=CC(=CN=C1)C(=O)N |
Specifications
98-92-0 | |
White | |
C6H6N2O | |
25 g | |
DFPAKSUCGFBDDF-UHFFFAOYSA-N | |
pyridine-3-carboxamide | |
936 | |
122.13 | |
Crystalline Powder |
129°C | |
6.0 to 7.5 (50g/L H2O soln.) | |
MFCD00006395 | |
nicotinamide, niacinamide, 3-pyridinecarboxamide, vitamin pp, nicotinic acid amide, papulex, aminicotin, amixicotyn, nicobion, nicotylamide | |
C1=CC(=CN=C1)C(=O)N | |
122.127 | |
CHEBI:17154 | |
≥99.0% (HPLC,T) | |
Nicotinamide |
Safety and Handling
EINECSNumber : (5)-0736
RTECSNumber : QS3675000
TSCA : Yes