missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Nile Red 99%

Supplier:  Thermo Scientific Chemicals 415711000

 View more versions of this product

Catalog No. AC415711000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nile Red
99%
98.5
99%
MFCD00011639
15, 6628
Solubility in water: almost insoluble. Other solubilities: soluble in acetone, acetonitrile, chloroform, thf,, ethanol, dmso, cs2, benzene, nitrobenzene, pyridin, e, hexane, diethyl ether
Authentic
9-(diethylamino)benzo[a]phenoxazin-5-one
65182
318.38
Pure
98.5% min.
Maroon to Red
Powder
203.0°C to 205.0°C
7385-67-3
100.0
C20H18N2O2
27, II, 468
Nile blue A oxazone
VOFUROIFQGPCGE-UHFFFAOYSA-N
CCN(CC)C1=CC=C2N=C3C(OC2=C1)=CC(=O)C1=CC=CC=C31
318.38
CHEBI:52169
99%
Glass bottle
Complies
100 mg

Chemical Identifiers

7385-67-3
318.38
VOFUROIFQGPCGE-UHFFFAOYSA-N
65182
CCN(CC)C1=CC=C2N=C3C(OC2=C1)=CC(=O)C1=CC=CC=C31
C20H18N2O2
MFCD00011639
Nile blue A oxazone
CHEBI:52169
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 230-966-0

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only