Nilfisk™ GD 10 Back Vacuum
Supplier: Nilfisk™ 9060709010
Description
- Lightweight, ergonomic design to reduce fatigue
- Low noise level for operator comfort and cleaning in noise-sensitive areas
- Upstream HEPA filter allows for safe collection of hazardous materials, trapping 99.99% of particles, down to and including 0.3 microns
- Standard equipped with a sack-style main filter that will prolong the life of both the HEPA filter and motor
- Exhaust air is directed upward so that it does not interfere with the operator
Specifications
Back Vacuum |