missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nimodipine, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215980325

 View more versions of this product

Catalog No. ICN15980325


Explore available promotions

Description

Description

Specifications

Specifications

Nimodipine
Yellow
MFCD00153848
nimodipine, nimotop, periplum, nimodipino, nimodipinum, nymalize, admon, nimodipinum inn-latin, nimodipino inn-spanish, isopropyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate
COCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)C
418.45
CHEBI:7575
Solid
66085-59-4
C21H26N2O7
25 mg
UIAGMCDKSXEBJQ-UHFFFAOYNA-N
3-(2-methoxyethyl) 5-propan-2-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
4497
418.4

Chemical Identifiers

66085-59-4
418.45
UIAGMCDKSXEBJQ-UHFFFAOYNA-N
4497
3-(2-methoxyethyl) 5-propan-2-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
C21H26N2O7
MFCD00153848
nimodipine, nimotop, periplum, nimodipino, nimodipinum, nymalize, admon, nimodipinum inn-latin, nimodipino inn-spanish, isopropyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate
CHEBI:7575
COCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)C
Videos
Safety and Handling

Safety and Handling

storageNote1 : Protect from light

Recommended Storage : Store at room temperature (15° to 30°C)

SDS
Documents

Documents

Promotions

Promotions