missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nimodipine, MP Biomedicals™
Supplier: MP Biomedicals Inc 0215980380
Specifications
Nimodipine | |
Yellow | |
MFCD00153848 | |
nimodipine, nimotop, periplum, nimodipino, nimodipinum, nymalize, admon, nimodipinum inn-latin, nimodipino inn-spanish, isopropyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate | |
COCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)C | |
418.45 | |
CHEBI:7575 | |
Solid |
66085-59-4 | |
C21H26N2O7 | |
100 mg | |
UIAGMCDKSXEBJQ-UHFFFAOYNA-N | |
3-(2-methoxyethyl) 5-propan-2-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate | |
4497 | |
418.4 |
Chemical Identifiers
66085-59-4 | |
418.45 | |
UIAGMCDKSXEBJQ-UHFFFAOYNA-N | |
4497 | |
3-(2-methoxyethyl) 5-propan-2-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate |
C21H26N2O7 | |
MFCD00153848 | |
nimodipine, nimotop, periplum, nimodipino, nimodipinum, nymalize, admon, nimodipinum inn-latin, nimodipino inn-spanish, isopropyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate | |
CHEBI:7575 | |
COCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)C |
Safety and Handling
storageNote1 : Protect from light
Recommended Storage : Store at room temperature (15° to 30°C)