Nine-Liter Wet Spirometer
Specially designed to meet the needs of today's physiology teachers.
Supplier: Customer Channels Group (Ccg) 7085301SU
Alert:
This item is intended for educational purposes only.
Description
This academic-quality wet spirometer combines reliability and accuracy with an increased volume to accommodate students with larger lung capacities.
- Large, easy-to-read scale graduations
- Includes disposable mouthpieces and a one-way plastic valve for sanitary classroom use
- Made of lightweight, yet durable, high-density polyethylene (HDPE) plastic and aluminum
Determine Five Different Respiratory Volumes:
- Tidal Volume (TV)
- Inspiratory Capacity (IC)
- Inspiratory Reserve Volume (IRV)
- Expiratory Reserve Volume (ERV)
- Vital Capacity (VC)
Specifications
Single-use | |
For measuring the volume of air inspired and expired by the lungs | |
Student wet spirometer |
Mouthpiece | |
100 Pack |