Nine-Liter Wet Spirometer
Specially designed to meet the needs of today's physiology teachers.
Supplier:  Customer Channels Group (Ccg) 7087200EMD
 View more versions of this product
Catalog No. S65066
Explore available promotions
Alert:
This item is intended for educational purposes only.
Description
Description
This academic-quality wet spirometer combines reliability and accuracy with an increased volume to accommodate students with larger lung capacities.
  • Large, easy-to-read scale graduations
  • Includes disposable mouthpieces and a one-way plastic valve for sanitary classroom use
  • Made of lightweight, yet durable, high-density polyethylene (HDPE) plastic and aluminum
Determine Five Different Respiratory Volumes:
  • Tidal Volume (TV)
  • Inspiratory Capacity (IC)
  • Inspiratory Reserve Volume (IRV)
  • Expiratory Reserve Volume (ERV)
  • Vital Capacity (VC)
Specifications
Specifications
Disposable mouthpieces, one-way plastic valve
For measuring the volume of air inspired and expired by the lungs
HDPE, Plastic and Aluminum
Student Wet Spirometer
Each