missing translation for 'onlineSavingsMsg'
Learn More

Ninhydrin, 99%

CAS: 485-47-2 | C9H6O4 | 178.143 g/mol

$48.71 - $699.05

Identifiants chimiques

CAS 485-47-2
Molecular Formula C9H6O4
Molecular Weight (g/mol) 178.143
MDL Number MFCD00003791
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
Synonym ninhydrin, ninhydrin hydrate, 1,2,3-indantrione monohydrate, ninhydrine, 2,2-dihydroxy-1h-indene-1,3 2h-dione, 2,2-dihydroxy-1,3-indandione, triketohydrindene hydrate, 1h-indene-1,3 2h-dione, 2,2-dihydroxy, 1,2,3-indantrione, 2-hydrate, 2,2-dihydroxyindane-1,3-dione
PubChem CID 10236
ChEBI CHEBI:86374
IUPAC Name 2,2-dihydroxyindene-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
voir les spécifications

Produits 4
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAA1040909
Afficher les documents
Thermo Scientific Chemicals
A1040909
10 g
chaque for $48.71
Il en reste null
 
AAA1040914
Afficher les documents
Thermo Scientific Chemicals
A1040914
25 g
chaque for $100.79
Il en reste null
 
AAA1040918
Afficher les documents
Thermo Scientific Chemicals
A1040918
50 g
chaque for $173.52
Il en reste null
 
AAA1040930
Afficher les documents
Thermo Scientific Chemicals
A1040930
250 g
chaque for $699.05
Il en reste null
 
Description

Description

Ninhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ninhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.

Solubility
Soluble in water, acetone, ethanol, and methanol. Slightly soluble in chloroform and ether.

Notes
Light Sensitive. Incompatible with strong oxidizing agents, amines, alkalis and strong bases.
Spécifications

Identifiants chimiques

485-47-2
178.143
FEMOMIGRRWSMCU-UHFFFAOYSA-N
10236
2,2-dihydroxyindene-1,3-dione
C9H6O4
MFCD00003791
ninhydrin, ninhydrin hydrate, 1,2,3-indantrione monohydrate, ninhydrine, 2,2-dihydroxy-1h-indene-1,3 2h-dione, 2,2-dihydroxy-1,3-indandione, triketohydrindene hydrate, 1h-indene-1,3 2h-dione, 2,2-dihydroxy, 1,2,3-indantrione, 2-hydrate, 2,2-dihydroxyindane-1,3-dione
CHEBI:86374
C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O

Spécifications

485-47-2
99%
MFCD00003791
1910963
14,6554
Soluble in water,acetone,ethanol,and methanol. Slightly soluble in chloroform and ether.
C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
178.143
CHEBI:86374
99%
Ninhydrin
251°C (decomposition)
C9H6O4
10 g
Light sensitive
ninhydrin, ninhydrin hydrate, 1,2,3-indantrione monohydrate, ninhydrine, 2,2-dihydroxy-1h-indene-1,3 2h-dione, 2,2-dihydroxy-1,3-indandione, triketohydrindene hydrate, 1h-indene-1,3 2h-dione, 2,2-dihydroxy, 1,2,3-indantrione, 2-hydrate, 2,2-dihydroxyindane-1,3-dione
FEMOMIGRRWSMCU-UHFFFAOYSA-N
2,2-dihydroxyindene-1,3-dione
10236
178.14
Powder
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H302-H315-H319-H335

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'einecsNumber' : 207-618-1

missing translation for 'rtecsNumber' : NK5425000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only