missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Ninhydrin, Reagent ACS

Supplier:  Thermo Scientific Chemicals 415721000

 View more versions of this product

Catalog No. AC415721000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ninhydrin
485-47-2
MFCD00003791
15, 6640
Solubility in water: soluble. Other solubilities: soluble in alcohol, slightly soluble in ether and chloroform
FEMOMIGRRWSMCU-UHFFFAOYSA-N
C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
178.143
CHEBI:86374
ACS Reagent
(to amino acids) Passes Test
Brown/White to White
Crystals
250.0°C
C9H6O4
01,732; 04,356
1, 2, 3-Indantrione, 1, 2, 3-Triketohydrindene, 2, 2-Dihydroxy-1H-indene-1, 3(2H)-dione, 1H-Indene-1, 2, 3-trione
Passes Test
Authentic
2,2-dihydroxyindene-1,3-dione
10236
178.14
Glass bottle
0.81
100 g

Chemical Identifiers

485-47-2
178.143
FEMOMIGRRWSMCU-UHFFFAOYSA-N
10236
2,2-dihydroxyindene-1,3-dione
C9H6O4
MFCD00003791
1, 2, 3-Indantrione, 1, 2, 3-Triketohydrindene, 2, 2-Dihydroxy-1H-indene-1, 3(2H)-dione, 1H-Indene-1, 2, 3-trione
CHEBI:86374
C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 207-618-1

RTECSNumber : NK5425000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only