missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Niobium(V) chloride, 99.8%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 201690500

 View more versions of this product

Catalog No. AC201690500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

254.0°C
204.7°C to 209.5°C
10026-12-7
99.75
Fine Crystalline Powder
99.75% min
NbCl5
14,214
niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratrem
YHBDIEWMOMLKOO-UHFFFAOYSA-I
pentachloroniobium
24818
99.8%
Niobium(V) chloride
99.8%
Yellow
100.0
50 g
Cl5Nb
MFCD00011127
15, 6643
Solubility in water: reacts with water. Other solubilities: soluble in hcl, soluble in ccl4, soluble in ethanol
Cl[Nb](Cl)(Cl)(Cl)Cl
270.16
270.16
Glass bottle

Chemical Identifiers

10026-12-7
270.16
YHBDIEWMOMLKOO-UHFFFAOYSA-I
24818
Cl[Nb](Cl)(Cl)(Cl)Cl
Cl5Nb
MFCD00011127
niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratrem
pentachloroniobium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 233-059-8

RTECSNumber : QU0350000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only