Learn More
Niobium(V) ethoxide, 99.99% (metals basis), Ta <500ppm, Thermo Scientific Chemicals
Preparation of niobium oxide and mixed metal oxides
$101.89 - $1097.53
Chemical Identifiers
CAS | 3236-82-6 |
---|---|
Molecular Formula | C10H25NbO5 |
Molecular Weight (g/mol) | 318.21 |
MDL Number | MFCD00015122 |
InChI Key | ZTILUDNICMILKJ-UHFFFAOYSA-N |
Synonym | niobium ethoxide, niobium 5+ ethanolate, ethanol, niobium 5+ salt, niobium 5+ ion pentakis ethoxide, pentaethoxyniobium, niobium pentaethoxide, ethanol, niobium 5+ salt 5:1, acmc-1cmht, niobium v pentaethoxide, ethanolate; niobium 5+ |
PubChem CID | 160675 |
IUPAC Name | ethanolate;niobium(5+) |
SMILES | CCO[Nb](OCC)(OCC)(OCC)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1468904
|
Thermo Scientific Chemicals
01468904 |
2 g |
Each for $101.89
|
|
|||||
AA1468909
|
Thermo Scientific Chemicals
01468909 |
10 g |
Each for $354.27
|
|
|||||
AA1468918
|
Thermo Scientific Chemicals
01468918 |
50 g |
Each for $1,097.53
|
|
|||||
Description
Preparation of niobium oxide and mixed metal oxides.Niobium(V) ethoxide is used for the sol-gel processing of materials containing niobium oxides and mixed metal oxides. It is also used to prepare ferroelectric nanomaterials consisting of oxides of potassium, silicon and niobium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3236-82-6 | |
318.21 | |
ZTILUDNICMILKJ-UHFFFAOYSA-N | |
160675 | |
CCO[Nb](OCC)(OCC)(OCC)OCC |
C10H25NbO5 | |
MFCD00015122 | |
niobium ethoxide, niobium 5+ ethanolate, ethanol, niobium 5+ salt, niobium 5+ ion pentakis ethoxide, pentaethoxyniobium, niobium pentaethoxide, ethanol, niobium 5+ salt 5:1, acmc-1cmht, niobium v pentaethoxide, ethanolate; niobium 5+ | |
ethanolate;niobium(5+) |
Specifications
3236-82-6 | |
1.258 g/mL | |
36°C (97°F) | |
1.516 | |
MFCD00015122 | |
UN2924 | |
Moisture sensitive | |
Soluble in organic solvents; Decomposes in water;Miscible with organic solvents. | |
CCO[Nb](OCC)(OCC)(OCC)OCC | |
318.21 | |
318.21 | |
Ta <500ppm | |
Niobium(V) ethoxide |
6°C | |
140°C to 142°C (0.1 mmHg) | |
C10H25NbO5 | |
Nb(OC2H5)5 | |
2 g | |
3678883 | |
niobium ethoxide, niobium 5+ ethanolate, ethanol, niobium 5+ salt, niobium 5+ ion pentakis ethoxide, pentaethoxyniobium, niobium pentaethoxide, ethanol, niobium 5+ salt 5:1, acmc-1cmht, niobium v pentaethoxide, ethanolate; niobium 5+ | |
ZTILUDNICMILKJ-UHFFFAOYSA-N | |
ethanolate;niobium(5+) | |
160675 | |
99.999% | |
Liquid | |
(metals basis) |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 221-795-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only