Learn More
Nisin, 900 I.U./mg, Thermo Scientific Chemicals
Nisin, CAS # 1414-45-5, is a polycyclic peptide derived from and produced by Lactococcus lactis that shows antibacterial activity against gram-positive bacteria.
Supplier: Thermo Scientific Chemicals J6637014
Description
Nisin, 900 I.U./mg is commonly used as a food preservative. It is also used as a selective agent in cell culture for the isolation of gram-negative bacteria, yeast, and molds. Nisin has been used in bioassays to study posttranslational modification. It binds and interferes with the permeability of the cytoplasmic membrane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nisin | |
900 I.U./mg | |
1414-45-5 | |
25 g | |
C143H230N42O37S7 | |
2967°C | |
Soluble in water | |
CCC(C)C1C(=O)NC(=C)C(=O)NC(C(=O)NC(CSCC(C(=O)N1)NC(=O)C(=CC)NC(=O)C(C(C)CC)N)C(=O)NC2C(SCC(NC(=O)CNC(=O)C3CCCN3C2=O)C(=O)NC(CCCCN)C(=O)NC4C(SCC(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC4=O)C)CC(C)C)CCSC)C(=O)NC(CC(=O)N)C(=O)NC(CCSC)C(=O)NC(CCCCN)C(=O)NC5C( | |
131847819 |
Nisin | |
Powder | |
Brown | |
>110°C (230°F) | |
MFCD00131724 | |
14,6564 | |
NVNLLIYOARQCIX-OAXMGIFASA-N | |
3354.09 | |
3354.06 |
Chemical Identifiers
1414-45-5 | |
3354.09 | |
NVNLLIYOARQCIX-OAXMGIFASA-N | |
CCC(C)C1C(=O)NC(=C)C(=O)NC(C(=O)NC(CSCC(C(=O)N1)NC(=O)C(=CC)NC(=O)C(C(C)CC)N)C(=O)NC2C(SCC(NC(=O)CNC(=O)C3CCCN3C2=O)C(=O)NC(CCCCN)C(=O)NC4C(SCC(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC4=O)C)CC(C)C)CCSC)C(=O)NC(CC(=O)N)C(=O)NC(CCSC)C(=O)NC(CCCCN)C(=O)NC5C( |
C143H230N42O37S7 | |
MFCD00131724 | |
131847819 |
Safety and Handling
EINECSNumber : 215-807-5
RTECSNumber : QU3340000
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only