missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nitric Acid 67-70%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™

Nitric Acid, HNO3, CAS Number-7697-37-2, acide nitrique, acidum nitricum, azotic acid, rfna, aqua fortis, nitryl hydroxide, salpetersaeure, nitrous fumes, nital, hydrogen nitrate, 500mL

Supplier:  Seastar Chemicals Inc S020101-TFIF03

 View more versions of this product

Catalog No. A467-500


Explore available promotions

Description

Description

Nitric Acid, HNO3, CAS Number-7697-37-2, acide nitrique, acidum nitricum, azotic acid, rfna, aqua fortis, nitryl hydroxide, salpetersaeure, nitrous fumes, nital, hydrogen nitrate, 500mL

  • Ideal for analysis by ICP-MS and high-resolution ICP-MS.
  • Evaluated for 65 metals at low-level ppt concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in specially manufactured and cleaned FEP fluoropolymer bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture - available on COA.
Specifications

Specifications

1
Colorless Yellow
7697-37-2
1.40 g/cm3
0.94kPa (20°C)
500 mL
Liquid
2031
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
O[N+]([O-])=O
63.01
CHEBI:48107
OPTIMA™
Nitric Acid
-41°C
120.5°C
FEP Bottle
Certificate of Lot Analysis is included with each bottle.
HNO3
MFCD00011349
15, 6663
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
63.01

Chemical Identifiers

7697-37-2
63.01
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid
HNO3
MFCD00011349
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
CHEBI:48107
O[N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8: Corrosive

SDS Documents

Documents

Accessories
Promotions

Promotions