Learn More
Nitric Acid (Certified ACS Plus), Fisher Chemical™
Nitric Acid material may darken during storage due to photochemical reaction
$403.31 - $3678.92
Chemical Identifiers
CAS | 7697-37-2 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
A200-500
|
Thermo Fisher Scientific
52006G500C |
500 mL | Glass Bottle |
Each for $403.31
Case of 6 Each for $2,419.86
|
|
||||
A200S-500
|
Thermo Fisher Scientific
FLA200S500 |
500 mL | Safe-Cote™ Glass |
Each for $474.62
Case of 6 Each for $2,847.72
|
|
||||
A200-212
|
Thermo Fisher Scientific
FLA200212 |
2.5 L | Glass Bottle |
Each for $671.46
|
|
||||
A200S-212
|
Thermo Fisher Scientific
FLA200S212 |
2.5 L | Safe-Cote™ Glass |
Case of 4 Each for $2,056.23
|
|
||||
A200-612GAL
|
Thermo Fisher Scientific
A200612GAL |
6.5 gal. | Glass Carboy |
Each for $3,678.92
|
|
||||
A200C-212
|
Thermo Fisher Scientific
FLA200C212 |
4 x 2.5 L | Glass Bottle |
Case of 4 Each for $1,748.54
|
|
||||
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
944 | |
nitric acid |
HNO3 | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
CHEBI:48107 | |
O[N+]([O-])=O |
Specifications
-1.0 | |
-41°C | |
120.5°C | |
0.2ppm max. | |
Glass Bottle | |
0.94 kPa at 20°C | |
70% | |
HNO3 | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
68.0 to 70.0% (w/w) | |
Nitric Acid |
Colorless Yellow | |
7697-37-2,7732-18-5 | |
1.40g/cm³ | |
Pass Test | |
2ppm max. | |
about 15.8 | |
500 mL | |
Liquid | |
2031 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
63.01 | |
Certified ACS Plus |
Safety and Handling
DANGER!
Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. May cause pulmonary edema. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.
NFPA
Health:4
Flammability:0
Instability:0
DOTInformation : DOT Class 8, : Corrosive