missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
![](http://beta-static.fishersci.ca/images/Acid.gif)
![](http://beta-static.fishersci.ca/images/Yellow.gif)
Nitric Acid (TraceMetal™ Grade), Fisher Chemical™
$445.92 - $905.25
Chemical Identifiers
CAS | 7697-37-2 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
A509P500
![]() |
Fisher Chemical
S010101TFIQ03 |
500 mL | HDPE Bottle |
Each for $445.92
|
![]() |
||||
A509P212
|
Fisher Chemical
S010101TFIQ09 |
2.5 L | HDPE Bottle |
Each for $905.25
|
|
||||
Description
Nitric Acid (TraceMetal™ Grade)
- Ideal for analysis by IC, GF or Flame AA, ICP-OES and ICP-MS.
- Evaluated for 65 metals at < 1 ppb concentrations.
- Manufactured,purified, and packaged in a Class 1000 clean room.
- Packaged in non-breakable polypropylene bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
- Two year shelf life from time of manufacture - available on COA. 45 mm bottle neck size
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
944 | |
nitric acid |
HNO3 | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
CHEBI:48107 | |
O[N+]([O-])=O |
Specifications
1.0 | |
-41°C | |
120.5°C | |
HDPE Bottle | |
Certificate of Lot Analysis included with each shipment. | |
HNO3 | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
67 to 70% (HNO3, w/w) | |
Nitric Acid |
Colorless Yellow | |
7697-37-2 | |
1.40g/cm³ | |
0.94 kPa at 20°C | |
500 mL | |
Liquid | |
2031 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
63.01 | |
TraceMetal |
Safety and Handling
![](http://beta-static.fishersci.ca/images/Acid.gif)
![](http://beta-static.fishersci.ca/images/Yellow.gif)
ShelfLife : 2 years
DOTInformation : DOT Class 8: Corrosive