missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nitrilotriacetic Acid (Certified ACS), Fisher BioReagents

$468.48 - $468.48

Chemical Identifiers

CAS 139-13-9
Molecular Formula C6H9NO6
Molecular Weight (g/mol) 191.14
MDL Number MFCD00004287
InChI Key MGFYIUFZLHCRTH-UHFFFAOYSA-N
Synonym nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
PubChem CID 8758
ChEBI CHEBI:44557
IUPAC Name 2-[bis(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CC(O)=O)CC(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP26701
SDS View Documents
Fisher BioReagents
BP26701
1 kg Poly Bottle
Each for $468.48
Case of 6 Each for $2,810.88
 
Description

Description

Nitrilotriacetic Acid is a chelating and sequestering agent. A builder in synthetic detergents.
Specifications

Chemical Identifiers

139-13-9
191.14
MGFYIUFZLHCRTH-UHFFFAOYSA-N
8758
2-[bis(carboxymethyl)amino]acetic acid
C6H9NO6
MFCD00004287
nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
CHEBI:44557
OC(=O)CN(CC(O)=O)CC(O)=O

Specifications

139-13-9
White
≥98.0 %
C6H9NO6
MFCD00004287
15, 6665
MGFYIUFZLHCRTH-UHFFFAOYSA-N
2-[bis(carboxymethyl)amino]acetic acid
8758
191.14
Certified ACS
Nitrilotriacetic Acid
245°C
2.3
Poly Bottle
N(CH2COOH)3
1 kg
nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
OC(=O)CN(CC(O)=O)CC(O)=O
191.14
CHEBI:44557
≥98.0%
Powder or Solid
Videos
Safety and Handling

Safety and Handling

Emergency Overview
Harmful if swallowed. Irritating to eyes and skin. Suspect cancer hazard.
NFPA
Health: 2

Signal Word: WARNING!

Recommended Storage : RT

SDS
missing translation for 'documents'

missing translation for 'documents'