missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nitrofurantoin, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2407936

 View more versions of this product

Catalog No. AAB2407936


Explore available promotions

Description

Description

Nitrofurantoin is a nitrofuran antibiotic that is used as a substrate of bacterial nitrofuran reductase to study the interactions of active metabolites with DNA, ribosomal proteins and metabolic and pro-oxidant processes. It is an antibactericidal compound that has been historically prepared by the reaction of 1-aminohydantoin sulfate and 5-nitro-2-furaldehyde diacetate. It shows activity against many Gram-positive and Gram-negative bacteria. Nitrofurantoin is effective against enterococci, staphylococci, streptococci, corneybacteria, many strains of Escherichia coli.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nitrofurantoin is a nitrofuran antibiotic that is used as a substrate of bacterial nitrofuran reductase to study the interactions of active metabolites with DNA, ribosomal proteins and metabolic and pro-oxidant processes. It is an antibactericidal compound that has been historically prepared by the reaction of 1-aminohydantoin sulfate and 5-nitro-2-furaldehyde diacetate. It shows activity against many Gram-positive and Gram-negative bacteria. Nitrofurantoin is effective against enterococci, staphylococci, streptococci, corneybacteria, many strains of Escherichia coli.

Solubility
Soluble in water (<0.01g/100mL at 19°C), dimethyl sulfoxide and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature. Incompatible with oxidizing agents. It is sensitive to light.
Specifications

Specifications

Nitrofurantoin
∼265°C (decomposition)
MFCD00003224
Hygroscopic; Light sensitive
Soluble in water (<0.01g/100 ml at 19°C),dimethyl sulfoxide and methanol.
[O-][N+](=O)C1=CC=C(O1)\C=N\N1CC(=O)NC1=O
238.16
CHEBI:71415
98%
67-20-9
C8H6N4O5
500 g
nitrofurantoin, macrodantin, 5-nitrofurantoin, furadantin, furadonine, nitrofurantoine, furadantine, furadantoin, furadoine, furadontin
NXFQHRVNIOXGAQ-YCRREMRBSA-N
1-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]imidazolidine-2,4-dione
6604200
238.16

Chemical Identifiers

67-20-9
238.16
NXFQHRVNIOXGAQ-YCRREMRBSA-N
6604200
1-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]imidazolidine-2,4-dione
C8H6N4O5
MFCD00003224
nitrofurantoin, macrodantin, 5-nitrofurantoin, furadantin, furadonine, nitrofurantoine, furadantine, furadantoin, furadoine, furadontin
CHEBI:71415
[O-][N+](=O)C1=CC=C(O1)\C=N\N1CC(=O)NC1=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H334-H341-H351-H361-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
May cause an allergic skin reaction.

P260-P264b-P270-P272-P280g-P285-P301+P312-P302+P352-P304+P341-P314-P330-P333+P313-P342+P311-P363-P501c

H302-H317-H334-H373

EINECSNumber : 200-646-5

RTECSNumber : MU2800000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only