Learn More
Nitromethane, 96%, for spectroscopy, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 167860025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopySpecifications
Nitromethane | |
75-52-5 | |
0.01 max. at 420nm, 0.05 max. at 405nm, 0.1 max. at 400nm, 0.2 max. at 395nm, 0.5 max. at 386nm, 1 max. at 380nm | |
-29°C | |
100°C to 102°C | |
95% min. (GC) | |
CH3NO2 | |
CH3NO2 | |
2.5 L | |
01,739; 04,357; 09,323; 13,199 | |
15, 6698 | |
Solubility in water: 95g/L (20°C). Other solubilities: soluble in alcohol,ether and dimethylformamide | |
C[N+]([O-])=O | |
61.04 | |
0.61 mPa.s (25°C) | |
61.04 | |
Spectroscopy |
96%, spectrophotometric grade | |
95.0 | |
100.0 | |
1.1200g/mL | |
35°C | |
Glass bottle | |
1.3805 to 1.3825 | |
MFCD00007400 | |
01, 74 | |
1.12 | |
methane, nitro, nitrocarbol, nitrometan, nitrometan polish, nitro-methane, unii-ru5wg8c3f4, ccris 1205, hsdb 106, meno2, ru5wg8c3f4 | |
LYGJENNIWJXYER-UHFFFAOYSA-N | |
nitromethane | |
6375 | |
CHEBI:77701 | |
96% | |
Liquid |
Chemical Identifiers
75-52-5 | |
61.04 | |
LYGJENNIWJXYER-UHFFFAOYSA-N | |
6375 | |
nitromethane |
CH3NO2 | |
MFCD00007400 | |
methane, nitro, nitrocarbol, nitrometan, nitrometan polish, nitro-methane, unii-ru5wg8c3f4, ccris 1205, hsdb 106, meno2, ru5wg8c3f4 | |
CHEBI:77701 | |
C[N+]([O-])=O |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Suspected of causing cancer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh
GHS Signal Word: Warning
EINECSNumber : 200-876-6
RTECSNumber : PA9800000
TSCA : TSCA
RUO – Research Use Only