Learn More
Nitronium hexafluorophosphate, Thermo Scientific Chemicals
$247.16 - $856.79
Chemical Identifiers
CAS | 19200-21-6 |
---|---|
Molecular Formula | F6NO2P |
Molecular Weight (g/mol) | 190.969 |
MDL Number | MFCD00031520 |
InChI Key | DXWZKTLGZWTYPL-UHFFFAOYSA-N |
Synonym | nitronium hexafluorophosphate, nitronium ion hexafluorophosphate, dioxidonitrogen 1+ hexafluorophosphate |
PubChem CID | 45933661 |
IUPAC Name | nitronium;hexafluorophosphate |
SMILES | [N+](=O)=O.F[P-](F)(F)(F)(F)F |
Description
Nitronium hexafluorophosphate is a nitrating reagent used in the nitration of alkanes such as methane, ethane, propane, n-butane, isobutane, neopentane and cyclohexane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
19200-21-6 | |
190.969 | |
DXWZKTLGZWTYPL-UHFFFAOYSA-N | |
45933661 | |
[N+](=O)=O.F[P-](F)(F)(F)(F)F |
F6NO2P | |
MFCD00031520 | |
nitronium hexafluorophosphate, nitronium ion hexafluorophosphate, dioxidonitrogen 1+ hexafluorophosphate | |
nitronium;hexafluorophosphate |
Specifications
White | |
Crystalline | |
F6NO2P | |
nitronium hexafluorophosphate, nitronium ion hexafluorophosphate, dioxidonitrogen 1+ hexafluorophosphate | |
DXWZKTLGZWTYPL-UHFFFAOYSA-N | |
nitronium;hexafluorophosphate | |
45933661 | |
Moisture sensitive | |
Nitronium hexafluorophosphate |
19200-21-6 | |
5 g | |
MFCD00031520 | |
Soluble in dichloromethane and nitroethane. | |
[N+](=O)=O.F[P-](F)(F)(F)(F)F | |
190.969 | |
190.97 | |
Off-white Cristals |
Safety and Handling
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : DOT Class: 8; Packaging Group: III
EINECSNumber : 242-872-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only