missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nitronium tetrafluoroborate, 0.5M solution in sulfolane, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 175130250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nitronium tetrafluoroborate | |
Colorless through Yellow | |
5.76,94.97 | |
25 mL | |
NO2BF4 | |
01,742; 04,358; 05,476; 08,361; 09,324; 06,520; 07,324; 09,414; 12,435; 13,274; 14,282; 17,314 | |
Solubility in water: reacts | |
[O-][NH+]=O.F[B-](F)(F)F | |
132.81 | |
132.81 | |
1.268 |
13826-86-3,126-33-0 | |
5.03,94.24 | |
Solution | |
BF4NO2 | |
MFCD00011432 | |
nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane | |
XYRQURYIJKJVSF-UHFFFAOYSA-N | |
nitronium;tetrafluoroborate | |
11073463 | |
Glass bottle | |
1.2680g/mL |
RUO – Research Use Only