missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitronium tetrafluoroborate, 97%
CAS: 13826-86-3 | BF4NO2 | 132.81 g/mol
$140.79 - $434.01
Chemical Identifiers
| CAS | 13826-86-3 |
|---|---|
| Molecular Formula | BF4NO2 |
| Molecular Weight (g/mol) | 132.81 |
| MDL Number | MFCD00011432 |
| InChI Key | XYRQURYIJKJVSF-UHFFFAOYSA-N |
| Synonym | nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane |
| PubChem CID | 11073463 |
| IUPAC Name | nitronium;tetrafluoroborate |
| SMILES | [O-][NH+]=O.F[B-](F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC174380050
|
Thermo Scientific Chemicals
174380050 |
5 g | Plastic bottle |
Each for $140.79
|
|
||||
|
AC174380250
|
Thermo Scientific Chemicals
174380250 |
25 g | Plastic bottle |
Each for $434.01
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13826-86-3 | |
| 132.81 | |
| XYRQURYIJKJVSF-UHFFFAOYSA-N | |
| 11073463 | |
| [O-][NH+]=O.F[B-](F)(F)F |
| BF4NO2 | |
| MFCD00011432 | |
| nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane | |
| nitronium;tetrafluoroborate |
Specifications
| 200.0°C | |
| 13826-86-3 | |
| 5 g | |
| BF4NO2 | |
| MFCD00011432 | |
| nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane | |
| XYRQURYIJKJVSF-UHFFFAOYSA-N | |
| nitronium;tetrafluoroborate | |
| 11073463 | |
| 97% | |
| Nitronium tetrafluoroborate |
| Colorless through White | |
| Adhering Crystals | |
| 96% min | |
| NO2BF4 | |
| 01,742; 04,358; 05,476; 08,361; 09,324; 01,747; 04,360; 07,253; 08,364; 14,215 | |
| Solubility in water: may decompose | |
| [O-][NH+]=O.F[B-](F)(F)F | |
| 132.81 | |
| 132.81 | |
| Plastic bottle |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 237-533-5
RUO – Research Use Only