missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nitrosobenzene 98.0+%, TCI America™
Supplier: TCI America N036725G
Specifications
Nitrosobenzene | |
66°C | |
57°C | |
MFCD00002059 | |
2811 | |
NLRKCXQQSUWLCH-UHFFFAOYSA-N | |
nitrosobenzene | |
11473 | |
107.11 | |
Crystalline Powder |
586-96-9 | |
White-Yellow | |
C6H5NO | |
25 g | |
benzene, nitroso, nitroso-benzene, unii-zi9w9e8g2z, nob, zi9w9e8g2z, chembl98797, nitroso benzene, keto-aniline, nitrosobenzene, acmc-209m6j | |
C1=CC=C(C=C1)N=O | |
107.112 | |
CHEBI:27986 | |
≥98.0% (GC) |
Chemical Identifiers
586-96-9 | |
107.112 | |
NLRKCXQQSUWLCH-UHFFFAOYSA-N | |
11473 | |
nitrosobenzene |
C6H5NO | |
MFCD00002059 | |
benzene, nitroso, nitroso-benzene, unii-zi9w9e8g2z, nob, zi9w9e8g2z, chembl98797, nitroso benzene, keto-aniline, nitrosobenzene, acmc-209m6j | |
CHEBI:27986 | |
C1=CC=C(C=C1)N=O |
Safety and Handling
EINECSNumber : (3)-2752
RTECSNumber : DA6497525
TSCA : Yes
Recommended Storage : Refrigerator