Learn More
Nitrosonium tetrafluoroborate, 98%, Thermo Scientific Chemicals
$82.86 - $705.71
Chemical Identifiers
CAS | 14635-75-7 |
---|---|
Molecular Formula | BF4HNO2 |
Molecular Weight (g/mol) | 133.82 |
MDL Number | MFCD00011433 |
InChI Key | XYRQURYIJKJVSF-UHFFFAOYSA-N |
Synonym | nitrosonium tetrafluoroborate, nitrosyl tetrafluoroborate, nitrosyltetrafluoroborate, nobf4, nitrilooxonium tetrafluoroborate, azanylidyneoxidanium tetrafluoroborate |
PubChem CID | 11137142 |
IUPAC Name | azanylidyneoxidanium;tetrafluoroborate |
SMILES | [O-][NH+]=O.F[B-](F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1580604
|
Thermo Scientific Chemicals
A1580604 |
2 g |
Each for $82.86
|
|
|||||
AAA1580609
|
Thermo Scientific Chemicals
A1580609 |
10 g |
Each for $188.32
|
|
|||||
AAA1580618
|
Thermo Scientific Chemicals
A1580618 |
50 g |
Each for $705.71
|
|
|||||
Description
Nitrosonium tetrafluoroborate is a nitrosating agent and oxidizer used in organic synthesis. It is also used to convert arylamines directly to diazonium tetrafluoroborates, which undergo the Balz-Schiemann reaction in situ to give high yields of aryl fluorides. Further, it is used as a catalyst for iodination of aromatics by iodide ion with air as oxidant, conversion of secondary amines to nitrosamines and in deamination of primary amides. It is involved in the conversion of arylhydrazines to azides. In addition to this, it reacts with ferrocene to prepare ferrocenium tetrafluoroborate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNitrosonium tetrafluoroborate is a nitrosating agent and oxidizer used in organic synthesis. It is also used to convert arylamines directly to diazonium tetrafluoroborates, which undergo the Balz-Schiemann reaction in situ to give high yields of aryl fluorides. Further, it is used as a catalyst for iodination of aromatics by iodide ion with air as oxidant, conversion of secondary amines to nitrosamines and in deamination of primary amides. It is involved in the conversion of arylhydrazines to azides. In addition to this, it reacts with ferrocene to prepare ferrocenium tetrafluoroborate.
Solubility
Soluble in acetonitrile.
Notes
Hygroscopic. Air and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids and amines.
Chemical Identifiers
14635-75-7 | |
133.82 | |
XYRQURYIJKJVSF-UHFFFAOYSA-N | |
11137142 | |
[O-][NH+]=O.F[B-](F)(F)F |
BF4HNO2 | |
MFCD00011433 | |
nitrosonium tetrafluoroborate, nitrosyl tetrafluoroborate, nitrosyltetrafluoroborate, nobf4, nitrilooxonium tetrafluoroborate, azanylidyneoxidanium tetrafluoroborate | |
azanylidyneoxidanium;tetrafluoroborate |
Specifications
250°C (0.01 mmHg)(Sublimation) | |
14635-75-7 | |
BF4HNO2 | |
UN3260 | |
nitrosonium tetrafluoroborate, nitrosyl tetrafluoroborate, nitrosyltetrafluoroborate, nobf4, nitrilooxonium tetrafluoroborate, azanylidyneoxidanium tetrafluoroborate | |
XYRQURYIJKJVSF-UHFFFAOYSA-N | |
azanylidyneoxidanium;tetrafluoroborate | |
11137142 | |
98% | |
Moisture sensitive | |
Nitrosonium tetrafluoroborate |
250°C (Sublimation) | |
2 g | |
MFCD00011433 | |
14,6649 | |
Soluble in acetonitrile. | |
[O-][NH+]=O.F[B-](F)(F)F | |
133.82 | |
116.82 | |
Irritating | |
2.185 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H318
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 238-679-2
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only