Chemglass Life Sciences NON MERCURY THERMOMETER DFS Item
Supplier:  Chemglass Inc CG350310
Thermometer, Non-Mercury, Partal Immersion, -20�C to 110�C, 305mm OAL, Thermometer is mineral spirit filled to eliminate workplace and environmental hazards associated with mercury. All ranges have 1� subdivisions. Thermometer has a white back and a 6 to 7mm outer diameter, and is calibrated for 76mm (3") immersion.
Catalog No. CG350310
Explore available promotions