Learn More
Henke-Sass, Wolf Non-Sterile 2-Part Syringes
These syringes feature a 2-part construction containing no rubber and eliminating the need for silicone oil. Made from polypropylene and polyethylene.
$59.24 - $4028.93
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance ISO 13485
Material Polyethylene (Plunger), Polypropylene (Barrel)
Color Clear
Latex Free Yes
View More Specs
Products 5
Catalog Number Mfr. No. Volume (Metric) Connection Type Includes Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Connection Type Includes Price Quantity  
14-817-183
View Documents
Henke-Sass, Wolf
AB1
1 mL Leur Slip 7000 Syringes
Case of 7000 Each for $4,028.93
Only null left
361046527
View Documents
Henke-Sass, Wolf
AB5LS
5 mL Leur Slip 3600 Syringes
Case of 3600 Each for $2,122.93
Only null left
14-817-188
View Documents
Henke-Sass, Wolf
AB5LL
5 mL Luer Lock 3600 Syringes
Case of 3600 Each for $1,976.91
Only null left
14-817-193
View Documents
Henke-Sass, Wolf
ABC5LS
5 mL Leur Slip 100 Syringes
Pack of 100 Each for $59.24
Case of 32 Pack for $1,895.68
Only null left
14-817-199
View Documents
Henke-Sass, Wolf
ABC20LL
20 mL Luer Lock 100 Syringes
Pack of 100 Each for $77.00
Case of 10 Pack for $770.00
Only null left
Description
Description
  • Disposable Syringes
  • 2-Part Syringe
  • Unsterile
  • Polypropylene Barrel
  • Polyethylene Plunger
  • Single-use
  • Latex Free
  • ISO
  • Chromatography
  • Laboratory Use
Specifications
Specifications
Not Autoclavable
Polyethylene (Plunger), Polypropylene (Barrel)
Yes
Bulk
Non-sterile
ISO 13485
Clear
Single-use
Yes
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'