Learn More
(+)-Nootkatone, crystalline, 98+%, Thermo Scientific Chemicals
$121.78 - $1759.70
Chemical Identifiers
CAS | 4674-50-4 |
---|---|
Molecular Formula | C15H22O |
Molecular Weight (g/mol) | 218.34 |
MDL Number | MFCD00036591 |
InChI Key | WTOYNNBCKUYIKC-SLEUVZQESA-N |
Synonym | unii-zms1vjk5hy, zms1vjk5hy, nootkatone,-, +-nootkatone, crystalline, 2 3h-naphthalenone, 4,4a,5,6,7,8-hexahydro-4,4a-dimethyl-6-1-methylethenyl-, 4s,4ar,6s, unii-3k3okv2a5a component, 4s,6s,4ar-4,4a-dimethyl-6-1-methylvinyl-3,4,5,6,7,8,4a-heptahydronaphthale n-2-one |
PubChem CID | 7567181 |
IUPAC Name | (4S,4aR,6S)-4,4a-dimethyl-6-prop-1-en-2-yl-3,4,5,6,7,8-hexahydronaphthalen-2-one |
SMILES | CC1CC(=O)C=C2C1(CC(CC2)C(=C)C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1916603
|
Thermo Scientific Chemicals
A1916603 |
1 g |
Each for $121.78
|
|
|||||
AAA1916606
|
Thermo Scientific Chemicals
A1916606 |
5 g |
Each for $460.47
|
|
|||||
AAA1916614
|
Thermo Scientific Chemicals
A1916614 |
25 g |
Each for $1,759.70
|
|
|||||
Description
(+)-Nootkatone, crystalline is used as an effective repellent in mosquitos, bed bugs, lone star ticks and head lice. It is also used as a food additive and flavoring material in beverages. It finds application in fragrances, cosmetics and pharmaceuticals. Further, it is used as a flavor material in beverage, frozen dairy and baked good applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(+)-Nootkatone, crystalline is used as an effective repellent in mosquitos, bed bugs, lone star ticks and head lice. It is also used as a food additive and flavoring material in beverages. It finds application in fragrances, cosmetics and pharmaceuticals. Further, it is used as a flavor material in beverage, frozen dairy and baked good applications.
Solubility
Slightly soluble in ethanol and chloroform. Partly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
4674-50-4 | |
218.34 | |
WTOYNNBCKUYIKC-SLEUVZQESA-N | |
7567181 | |
CC1CC(=O)C=C2C1(CC(CC2)C(=C)C)C |
C15H22O | |
MFCD00036591 | |
unii-zms1vjk5hy, zms1vjk5hy, nootkatone,-, +-nootkatone, crystalline, 2 3h-naphthalenone, 4,4a,5,6,7,8-hexahydro-4,4a-dimethyl-6-1-methylethenyl-, 4s,4ar,6s, unii-3k3okv2a5a component, 4s,6s,4ar-4,4a-dimethyl-6-1-methylvinyl-3,4,5,6,7,8,4a-heptahydronaphthale n-2-one | |
(4S,4aR,6S)-4,4a-dimethyl-6-prop-1-en-2-yl-3,4,5,6,7,8-hexahydronaphthalen-2-one |
Specifications
4674-50-4 | |
0.997 | |
99°C (210°F) | |
MFCD00036591 | |
4676969 | |
Slightly soluble in ethanol and chloroform. Partly soluble in water. | |
+185° (c=1 in Chloroform) | |
(4S,4aR,6S)-4,4a-dimethyl-6-prop-1-en-2-yl-3,4,5,6,7,8-hexahydronaphthalen-2-one | |
7567181 | |
≥98% | |
(+)-Nootkatone |
34°C to 36°C | |
125°C (0.5 mmHg) | |
C15H22O | |
1 g | |
unii-zms1vjk5hy, zms1vjk5hy, nootkatone,-, +-nootkatone, crystalline, 2 3h-naphthalenone, 4,4a,5,6,7,8-hexahydro-4,4a-dimethyl-6-1-methylethenyl-, 4s,4ar,6s, unii-3k3okv2a5a component, 4s,6s,4ar-4,4a-dimethyl-6-1-methylvinyl-3,4,5,6,7,8,4a-heptahydronaphthale n-2-one | |
WTOYNNBCKUYIKC-SLEUVZQESA-N | |
CC1CC(=O)C=C2C1(CC(CC2)C(=C)C)C | |
218.34 | |
218.34 | |
Crystalline |
Safety and Handling
EINECSNumber : 225-124-4
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only