missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Norethindrone, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215594683

 View more versions of this product

Catalog No. ICN15594683

Explore available promotions

Description

Description

Specifications

Specifications

Norethindrone
White
0.25 g
VIKNJXKGJWUCNN-XGXHKTLJSA-N
(8R,9S,10R,13S,14S,17R)-17-ethynyl-17-hydroxy-13-methyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
6230
298.4
68-22-4
C20H26O2
norethindrone, norethisterone, norethisteron, micronor, 19-norethisterone, norcolut, noriday, norlutin, primolut-n, norethynodrone
CC12CCC3C(C1CCC2(C#C)O)CCC4=CC(=O)CCC34
298.426
CHEBI:7627
Powder

Chemical Identifiers

68-22-4
298.426
norethindrone, norethisterone, norethisteron, micronor, 19-norethisterone, norcolut, noriday, norlutin, primolut-n, norethynodrone
CHEBI:7627
CC12CCC3C(C1CCC2(C#C)O)CCC4=CC(=O)CCC34
C20H26O2
VIKNJXKGJWUCNN-XGXHKTLJSA-N
6230
(8R,9S,10R,13S,14S,17R)-17-ethynyl-17-hydroxy-13-methyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
Videos
SDS
Documents

Documents

Promotions

Promotions