Honeywell™ North™ A700 Series Safety Glasses
Sporty styling provides fashionable fit for the everyday wearer
Supplier:  Honeywell™ A702
 View more versions of this product
Catalog No. 19-160-0219
Explore available promotions
Description
Description
  • Nine-base polycarbonate hardcoated lenses provide 180° field of distortion-free vision; block 99.9% UV
  • Lightweight design with sport temples and soft temple-tip pads for secure, wraparound fit
  • Molded universal nose bridge for comfortable fit
Compliance
Meet or exceed ANSI™ Z87.1-2010 standards.
Specifications
Specifications
Meets or exceed ANSI Z87.1-2003 standards.
Amber
Clear
Polycarbonate