Honeywell™ North™ Quantitative Fit-Test Adapter
Measures fit of the actual respirator a worker wears on the job
Supplier: Honeywell™ 770021
Description
- Eliminates need to inventory several sizes of probed models for fit-testing
- To use, just thread the adapter onto one of the facepiece's cartridge connectors
- Attach a matching cartridge or filter onto the other cartridge connector
- Respirator is then ready to be connected to a quantitative fit-testing apparatus.
- Use to fit-test North 5400, 5500, 7600, and 7700 Series respirators
- For airline and SCBA facepieces, use the appropriate facepiece converter in conjunction with the adapter (converter not required for CF2000 Series Continuous Flow Airline Respirator)
Specifications
Fit Test Adapter | |
Eliminates need to inventory several sizes of probed models for fit-testing | |
5400, 5500, 7600 and 7700 series respirators, portacount™ quantitative fit tester | |
1 Ea. |