Honeywell™ North™ Series 7900 Emergency Escape Mouthbit Respirator
The 7900 Series mouthbit respirators offer a quick and economical method to escape non-IDLH emergency situations. Use the respirator as a visitor escape device or issue individual respirators to employees as a safety precaution against a potential chemical leak
$40.93 - $40.93
Specifications
Certifications/Compliance NIOSH
Product Type Emergency Escape Mouthpiece Respirator
For Use With (Application) Emergency use at pulp and paper mills, water treatment plants, pools and spas, chemical plants and other facilities
Includes Single cartridge, mouthpiece, neck strap and nose clip
Size Universal
View More Specs
Products 1
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
19-816-212
View Documents
Honeywell™
7902
Emergency Escape Mouthpiece Respirator
Each for $40.93
Case of 10 Each for $379.20
Description
Description
  • Provides escape from acid gas or ammonia
  • Convenient neckstrap or beltclip
  • Lightweight and disposable
Model 7902 is designed for “escape only” from atmospheres containing chlorine, chlorine dioxide, sulfur dioxide, and hydrogen sulfide. Use Model 7904 for environments where ammonia is present.
Low breathing resistance increases wearer acceptance and reduces fatigue. Exhalation valve prevents inward contamination leakage; inhalation valve keeps exhaled breath and spittle from contacting chemical absorbent area. Break-away neck strap reduces risk of injury should respirator become caught in machinery. Large stainless-steel belt clip ensures easy attachment to belt or pants.
Specifications
Specifications
NIOSH
Emergency use at pulp and paper mills, water treatment plants, pools and spas, chemical plants and other facilities
Universal
Emergency Escape Mouthpiece Respirator
Single cartridge, mouthpiece, neck strap and nose clip
SDS
missing translation for 'documents'
missing translation for 'documents'