Honeywell North™ North Zone Ratchet Cap Style Hard Hat
Contemporary shell design with four large areas for custom logo imprinting or other personalization options
Supplier:  Honeywell North™ N10R020000
 View more versions of this product
Catalog No. 17378697
Explore available promotions
Description
Description
  • 3-level height adjustment (low, medium, high) for greater stability, balance and retention
  • Removable
    • Replaceable rear comfort cradle for improved fit and comfort
  • Easy-grip ratchet for smooth and precise headband adjustment
    • Even when wearing gloves
  • Ergonomic headband for adjustment to 3 different sizes (small, medium/large and X-large)
  • Moisture-wicking, breathable sweatband
    • Easily removeable, washable and replaceable
  • Short brim and low-profile design for increased field of vision and user mobility
  • Slots for cap-mounted earmuffs, face shields and other PPE accessories
Specifications
Specifications
ANSI/ISEA Z89.1 2014: Type I, Class C, G, and E, CSA Z94.1 2015: Type I, Class C, G, and E
Short Brim Hard Hat
Not Ventilated
21.84 cm
Yellow
Universal
Short Brim
8.6 in.