MilliporeSigma™ Novagen™ ColiRollers™ Plating Beads
Used for plating bacteria for transformation molecular cloning applications.
Supplier:  MilliporeSigma™ 710133
 View more versions of this product
Catalog No. 710133
Explore available promotions
Description
Description
EMD Millipore Novagen™ ColiRollers™ Plating Beads provides even distribution of colonies.
  • Specially treated glass beads
  • Eliminates inconsistencies of conventional methods
  • Provides even distribution of colonies
  • Sterile
  • Each package contains beads for 60 to 80 plates
  • Packaging: Glass Bottle
Specifications
Specifications
15°C to 30°C
1 Pack
Solid