missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Novagen™ CytoBuster™ Protein Extraction Reagent

Formulation of detergents optimized for efficient extraction of soluble proteins from mammalian and insect cells

Supplier:  MilliporeSigma™ MIL710094

 View more versions of this product

Catalog No. 710094MI


Explore available promotions

Description

Description

  • Gentle, non-ionic composition of reagent enables isolation of functionally active endogenous or expressed proteins without need for secondary treatment such as sonication or freeze/thaw
  • Formulated for utilization in western blotting, immunoprecipitation and kinase/phosphatase assays
  • Reagent is compatible with protease inhibitors, kinase inhibitors and phosphatase inhibitors

Applications:

Western blotting, immunoprecipitation and kinase/phosphatase assays

Specifications

Specifications

250mL
Protein Extraction Reagent
Western blotting, immunoprecipitation and kinase/phosphatase assays
15° to 30°C
Videos
SDS
Documents

Documents

Promotions

Promotions