missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Novagen™ Pellet Paint™ Co-Precipitant
Compatible with most molecular biology procedures and is free of contaminating nucleic acids and nucleolytic enzymes
$234.00 - $1260.00
Specifications
| Content And Storage | Store at −20°C |
|---|---|
| Label or Dye | Fluorescent dye-labeled |
| Form | Liquid |
Description
Compatible with most molecular biology procedures and is free of contaminating nucleic acids and nucleolytic enzymes
Direct visualization and tracking of precipitated materialSpecifications
| Store at −20°C | |
| Fluorescent dye-labeled | |
| Liquid |