missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ 10X Bolt™ Sample Reducing Agent
Optimized for use with Bolt™ Bis-Tris Plus gels, and available in a variety of formats.
Supplier: Invitrogen™ B0004
Description
- Bolt™ Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of small- to medium-sized proteins under denaturing conditions
- Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications
- Bolt™ gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial
- Also use Bolt™ gels to obtain optimal results for day-to-day protein separation needs
- Gels come in four gel types and multiple well formats
- A Mini Gel Tank is required for use of all Bolt™ gels
- Run times may vary depending upon the type of running buffer used
- Store at 2°C to 8°C
Specifications
Sample Reducing Agent (10X) contains 500 mM dithiothreitol (DTT). Store at 2°C to 8°C. |
|
Bolt Mini Gel Tank | |
Liquid | |
Bolt™ Bis-Tris Plus Gels | |
250 μL |
10X, 500 mM | |
Sample Reducing Agent | |
500 mM dithiothreitol (DTT) | |
Bolt™ | |
Reducing Agent |
For Research Use Only. Not for use in diagnostic procedures.