Learn More
Invitrogen™ 4X Bolt™ LDS Sample Buffer
Optimized for use with Bolt™ Bis-Tris Plus gels, and are available in a variety of formats
$29.69 - $230.00
Specifications
Content And Storage | Sample Buffer (4X) containing lithium dodecyl sulfate (LDS) at pH 8.5 with SERVA Blue G250 and phenol red. Store at 4°C to 25°C. |
---|---|
Buffer | Sample Loading Buffers |
Concentration | 4X |
For Use With (Equipment) | Bolt Mini Gel Tank |
Gel Type | Bis-Tris Plus |
Includes: lithium dodecyl sulfate (LDS) at pH 8.4.
Description
Novex™ Bolt™ buffers and sample reducing agents are optimized for use with Bolt Bis-Tris Plus gels, and are available in a variety of formats.
About Bolt™ Bis-Tris Plus Gels
Bolt™ Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of your small- to medium-sized proteins under denaturing conditions. Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt™ gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt™ gels to obtain optimal results for your day-to-day protein separation needs. Bolt™ Bis-Tris Plus gels come in four gel types and multiple well formats. A Mini Gel Tank is required for use of all Bolt™ gels.
Bolt™ gel run times may vary depending upon the type of running buffer used.
Specifications
Sample Buffer (4X) containing lithium dodecyl sulfate (LDS) at pH 8.5 with SERVA Blue G250 and phenol red. Store at 4°C to 25°C. |
|
4X | |
Bis-Tris Plus | |
Bolt™ Bis-Tris Plus Gels |
Sample Loading Buffers | |
Bolt Mini Gel Tank | |
LDS Sample Buffer | |
Bolt™ |
For Research Use Only. Not for use in diagnostic procedures.