missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ ELISA Buffer Kit DFS Item

Concentrated

Supplier:  Invitrogen™ CNB0011

Catalog No. CNB0011


Explore available promotions

Includes: Coating Buffer A (100mL): 1 bottle, Coating Buffer B (100mL): 1 bottle, Assay Buffer (5x) (200mL): 1 bottle, Wash Buffer (25x) (100mL): 3 bottles, Stabilized Chromogen (25mL): 4 bottles, Stop Solution (100mL): 1 bottle.

Description

Description

Buffer Kit for Antibody Pairs is for use with Invitrogen Antibody Pairs. Kit provides sufficient materials for ten ELISA plates.

Build Your Own Immunoassay, Cell Analysis, Immunoassays

Order Info

Shipping Condition: Wet Ice

Specifications

Specifications

10 plates
ELISA
Wet Ice
Concentrate
Coating Buffer A (100 mL): 1 bottle
Coating Buffer B (100 mL): 1 bottle
Assay Buffer (5x) (200 mL): 1 bottle
Wash Buffer (25x) (100 mL): 3 bottles
Stabilized Chromogen (25 mL): 4 bottles
Stop Solution (100 mL): 1 bottle. Store at 2°C to 8°C.
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.