Learn More
Invitrogen™ Novex™ Hi-Density TBE Sample Buffer (5X)
Compatible with polyacrylamide gels
Supplier: Invitrogen™ LC6678
Description
Novex TBE Gels provide the highest resolution possible for analyzing restriction digests and PCR products. DNA fragments ranging from 10 to 3,000 base pairs are clearly resolved into sharp, tight bands. Novex DNA Retardation gels are prepared with 0.5X TBE as the gel buffer. This is sufficient for good electrophoretic separation, yet low enough to promote DNA-protein interactions
- Novex TBE and DNA Retardation Gels are designed to run on XCell SureLock™ Mini-Cell
- Formulation: Novex TBE and DNA Retardation Gels are manufactured with high-purity tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, and APS
- Recommended Buffers: For optimum performance, Novex TBE Running Buffer and Novex Hi-Density TBE Sample Buffer are strongly recommended for use with these gels
- Novex Hi-Density TBE Sample Buffer contains the density agent Ficoll™, which yields sharper, straighter bands than conventional density agents, and tracking dyes Bromophenol Blue and Xylene Cyanol
Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting
Order Info
Shipping Condition: Wet Ice
Specifications
| Sample Loading Buffer | |
| Store at 2°C to 8°C | |
| Nucleic Acid Gel Electrophoresis, Blotting | |
| Novex | |
| 10 mL |
| 5 X | |
| Polyacrylamide Gels | |
| Bromophenol Blue, Xylene Cyanol | |
| TBE Buffer | |
| Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.