missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ HiMark™ Pre-stained Protein Standard DFS Item

Designed for analysis of high molecular weight proteins on NuPAGE™ Novex™ Tris-Acetate gels

Supplier:  Invitrogen™ LC5699

Catalog No. LC5699


Explore available promotions

Includes: 250μL of HiMark™ Pre-Stained HMW Protein Standard; Storage Buffer: Tris-HCl, Formamide, SDS, Phenol Red

Description

Description

  • Comprehensive - contains markers with a wide range of high molecular weights, providing nine protein bands in the range of 30 to 460kDa
  • Convenient - a ready-to-use formulation eliminates the need to heat, reduce, or add sample buffer prior to use
  • Efficient - allows easy visualization of electrophoresis and rapid evaluation of Western transfer efficiency
  • Compatible - intended for use with NuPAGE Novex 3 to 8% and 7% Tris-Acetate gels under denaturing conditions
  • Can also be used with NuPAGE Novex 4-12% Bis-Tris Gels with NuPAGE MOPS Running Buffer and Novex 4% Tris-Glycine Gel, however to obtain best results with HMW proteins, always use NuPAGE Novex Tris-Acetate Gels
  • Gel Compatibility: NovexTris-Glycine Gels, NuPAGE Tris-Acetate Gels

HiMark™ High Molecular Weight Protein Standard also available in an unstained format

1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Dry Ice

Specifications

Specifications

Contents: 250 μL of HiMark™ Pre-Stained HMW Protein Standard
Storage Buffer: Tris-HCl, Formamide, SDS, Phenol Red
Storage:Store at -20°C
9
Yes
30 to 460 kDa
Novex™ Tris-Glycine Gels, NuPAGE™ Tris-Acetate Gels
HiMark™
Dry Ice
Colorimetric
Protein Ladder
Tris-acetate gels
2 colors: Blue, Pink
460, 268, 238, 171, 117, 71, 55, 41, 31 kDa
250 μL
Western Blotting, SDS-PAGE
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.