Learn More
Invitrogen™ Human IL-6 ELISA Kit
ELISA
Supplier: Invitrogen™ KHC0061
Description
The Human Interleukin 6 (Hu IL-6) ELISA quantitates Hu IL-6 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-6. Principle of the method The Human IL-6 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
IL-6 is a gene that encodes a cytokine that plays a role in inflammation and the maturation of B cells. The protein produced by this gene is an endogenous pyrogen that can induce fever in people with autoimmune diseases or infections. The gene is primarily expressed at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis. Elevated levels of the encoded protein have been found in virus infections, including COVID-19 (disease caused by SARS-CoV-2). Diseases associated with IL6 include Kaposi Sarcoma and Rheumatoid Arthritis, Systemic Juvenile.Specifications
P05231 | |
<2 pg/mL | |
ELISA | |
Human | |
Colorimetric Microplate Reader | |
BSF-2,BSF2,CDF,HGF,HSF,IFN-beta-2,IFNB2,IL-6 | |
6.2 | |
HRP | |
96 Tests | |
Plasma 100 μL, Serum 100 μL, Supernatant 100 μL | |
IL-6 | |
3 hrs |
7.8 to 2500 pg/mL | |
HRP | |
Plasma, Serum, Supernatant | |
Colorimetric | |
3569 | |
7.9% | |
Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
<2 pg/mL | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
For Research Use Only. Not for use in diagnostic procedures.