missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ Sharp Unstained Protein Standard DFS Item

Designed for accurate, easy and convenient molecular weight estimation of a wide range of molecular weight proteins during SDS-PAGE and western blotting

Supplier:  Invitrogen™ LC5801

Catalog No. LC5801


Explore available promotions

Description

Description

  • Accurate - Sharp bands for accurate estimation of molecular weigh
  • Comprehensive - A wide range of molecular weight bands consisting of 12 proteins in the range of 3.5 - 260kDa
  • Convenient - A ready-to-use formulation eliminates the need to heat, reduce, or add sample buffer prior to use
  • Adaptable - Suitable for most gel types, recommended for use with Novex™ NuPAGE™, Tris-Glycine, and Tricine Gels
  • Compatible with common gel stains - Works with SimplyBlue™, Coomassie™, silver and SYPRO™ staining method
  • Gel Compatibility: NovexTricine Gels, NovexTris-Glycine Gels, NuPAGE Bis-Tris Gels, NuPAGE Tris-Acetate Gels, SDS-PAGE Gels

The Novex Sharp Protein Standard is also available in a pre-stained format.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Dry Ice

Specifications

Specifications

Contents: two vials of 250 μL each

Storage buffer: 65 mM Tris (pH 6.8), 30% glycerol, 2% SDS, 2.5 mM EDTA, 0.0025% Coomassie G-250

Storage: Upon receipt store at -20°C

Protein Ladder
3.5 to 260 kDa
Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Novex™
Dry Ice
12
Yes
Unstained
260, 160, 110, 80, 60, 50, 40, 30, 20, 15, 10, 3.5 kDa
2 x 250 μL
Western Blotting, SDS-PAGE
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.